Benzoic acid, Certipur® Reag. USP, Reag. Ph. Eur. volumetric standard, secondary reference material for alkalimetry, traceable to NIST Standard Reference Material (SRM), Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.02401.0060
EM1.02401.0060CS 14918 USD
EM1.02401.0060
Benzoic acid, Certipur® Reag. USP, Reag. Ph. Eur. volumetric standard, secondary reference material for alkalimetry, traceable to NIST Standard Reference Material (SRM), Supelco®
Benzoic acid

Merck have introduced into the range of Certipur® standards some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: C₇H₆O₂
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 121…123 °C
Density: 1.316 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
ADR: 8,II
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

N/A N/A


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR