Calcium chloride tetrahydrate, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.02384.0100
EM1.02384.0100CS 61253 USD
EM1.02384.0100
Calcium chloride tetrahydrate, Suprapur® for trace metal analysis, Supelco®
Calcium chloride tetrahydrate
Formula: CaCl₂·4H₂O
MW: 183.04 g/mol
Boiling Pt: ∼1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 1.71 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 25094-02-4
EINECS: 233-140-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 772 °C (anhydrous substance)
Molar Mass 183.05 g/mol
pH 5 - 7 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR