Calcium sulfate dihydrate, EMSURE® for analysis, precipitated, Supelco®

Supplier: MilliporeSigma

1.02161.0500
EM1.02161.0500CS 770 USD
EM1.02161.0500
Calcium sulfate dihydrate, EMSURE® for analysis, precipitated, Supelco®
Calcium sulphate dihydrate
Formula: CaSO₄·2H₂O
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Molar Mass 172.17 g/mol
Bulk Density 400 - 600 kg/m3
Density 2.32 g/cm3 (20 °C)
pH 7.0 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR