Calcium hydroxide for the preparation of DIN 19266 pH standard buffer solutions, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Slaked lime

1.02111.0025
EM1.02111.0025CS 9384 USD
EM1.02111.0025
Calcium hydroxide for the preparation of DIN 19266 pH standard buffer solutions, Supelco®
Calcium hydroxide
Formula: Ca(OH)₂
MW: 74.09 g/mol
Melting Pt: 550 °C
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III
Merck Index: 13,01675

Order Now

Specification Test Results

Solubility 1.7 g/l (20 °C)
Melting Point 550 °C (decomposition)
Molar Mass 74.09 g/mol
Bulk Density 400 kg/m3
Boiling Point 2850 °C (1013 hPa) (decomposition)
Vapor Pressure (20 °C)
Density 2.24 g/cm3 (20 °C)
pH 12.6 (H2O, 20 °C) (saturated solution)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR