Cesium chloride, EMSURE®, Extra Pure for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.02041.1000
EM1.02041.1000CS 44712 USD
EM1.02041.1000
Cesium chloride, EMSURE®, Extra Pure for analysis, Supelco®
Cesium chloride
Formula: CsCl
MDL Number: MFCD00010955
CAS Number: 7647-17-8

Order Now

Specification Test Results

Solubility 1860 g/l (20 °C)
Melting Point 646 °C
Molar Mass 168.36 g/mol
Bulk Density 1800 kg/m3
Boiling Point 1382 °C
Density 3.97 g/cm3 (20 °C)
pH 7 - 9 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR