Cadmium, granular, EMSURE® ∼ 3-6 mm for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.02004.0250
EM1.02004.0250CS 53269 USD
EM1.02004.0250
Cadmium, granular, EMSURE® ∼ 3-6 mm for analysis, Supelco®
Cadmium
Formula: Cd
MW: 112.41 g/mol
Boiling Pt: 765
Melting Pt: 321
Density: 8.6 g/cm3 (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 3288
ADR: 6.1 III
Merck Index: 12,01649

Order Now

Specification Test Results

Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.001 %
Hg (Mercury) ≤ 0.0005 %
Pb (Lead) ≤ 0.005 %
Zn (Zinc) ≤ 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR