1-Butanol, LiChrosolv® for liquid chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol

M-Clarity™ quality level = MQ100

1.01988.1000 1.01988.2500
EM1.01988.1000CS 1384 USD
EM1.01988.1000 EM1.01988.2500
1-Butanol, LiChrosolv® for liquid chromatography, Supelco®
1-Butanol
Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: -89
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 30
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR