1-Butanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol
M-Clarity™ quality level = MQ100
1.01988.1000
1.01988.2500
EM1.01988.1000CS
1384
USD
EM1.01988.1000
EM1.01988.2500
1-Butanol, LiChrosolv® for liquid chromatography, Supelco®
1-Butanol
Formula:
H₃C(CH₂)₂CH₂OH MW: 74.12 g/mol Boiling Pt: 117.6 Melting Pt: -89 Density: 0.812 g/cm³ (20 °C) Flash Pt: 30 Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3 III REACH: 01-2119484630-38 Merck Index: 13,01539 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...