tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: MTBE, 2-Methyl-2-methoxypropane, Ether, tert-butyl methyl, Methyl 1,1-dimethylethyl ether, 2-Methoxy-2-methylpropane, Methyl t-butyl ether, Methyl tert-butyl ether, Methyl-t-Butyl Ether

1.01984.1000
EM1.01984.1000CS 1619 USD
EM1.01984.1000
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
tert-Butyl methyl ether

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.2 °C (1013 hPa)
Melting Pt: –108.6 °C
Density: 0.7405 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 42 g/l (20 °C)
Melting Point -108.6 °C
Molar Mass 88.15 g/mol
Boiling Point 55.3 °C (1013 hPa)
Vapor Pressure 268 hPa (20 °C)
Flash Point -28 °C
Explosion Limit 1.5 - 8.5 %(V)
Density 0.74 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 460 °C
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR