Potassium hydrogen phthalate, Certipur® certified secondary standard reference material for pH measurement, directly traceable to primary reference material (SRM) from NIST/PTB, Supelco®

Supplier: MilliporeSigma

Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt

1.01965.0025
EM1.01965.0025CS 10477 USD
EM1.01965.0025
Potassium hydrogen phthalate, Certipur® certified secondary standard reference material for pH measurement, directly traceable to primary reference material (SRM) from NIST/PTB, Supelco®
Potassium hydrogen phthalate
Formula: HOOCC₆H₄COOK
Melting Pt: 295…300 °C
Density: 1.6362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 80 g/l (20 °C)
Melting Point 295 - 300 °C
Molar Mass 204.22 g/mol
Bulk Density 900 kg/m3
Density 1.636 g/cm3
pH 4.0 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR