di-Sodium tetraborate decahydrate, Certipur® certified secondary standard reference material for pH measurement, directly traceable to primary reference material (SRM) from NIST/PTB, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

1.01964.0025
EM1.01964.0025CS 10477 USD
EM1.01964.0025
di-Sodium tetraborate decahydrate, Certipur® certified secondary standard reference material for pH measurement, directly traceable to primary reference material (SRM) from NIST/PTB, Supelco®
di-Sodium tetraborate decahydrate
Formula: Na₂B₄O₇·10H₂O
MW: 381.37 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662

Order Now

Specification Test Results

Solubility 51.4 g/l (20 °C)
Melting Point 75 °C Elimination of water of crystallisation
Molar Mass 381.32 g/mol
Bulk Density 750 kg/m3
Vapor Pressure 0.213 hPa (20 °C)
Density 1.72 g/cm3 (20 °C)
pH 9.2 (47 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR