Sodium hydrogen carbonate/sodium carbonate certified secondary standard reference material for pH measurement; directly traceable to primary SRM from NIST/PTB pH(S)=10.014 (25°C) (DIN 19266) Certipur®
Supplier: MilliporeSigma
1.01962.0001
EM1.01962.0001CS
14906
USD
EM1.01962.0001
Sodium hydrogen carbonate/sodium carbonate certified secondary standard reference material for pH measurement; directly traceable to primary SRM from NIST/PTB pH(S)=10.014 (25°C) (DIN 19266) Certipur®
Buffers
pH Calibration Buffers