1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate indicator for metal titration, Millipore®

Supplier: MilliporeSigma
Warning

Synonyms: Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulfonate monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate, Catechol-3,5-disulfonic acid disodium salt monohydrate, 3,5-Pyrocatecholdisulfonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate

1.01922.0100
EM1.01922.0100CS 9083 USD
EM1.01922.0100
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate indicator for metal titration, Millipore®
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate
Formula: C₆H₄Na₂O₈S₂·1H₂O
MDL Number: MFCD00149531
CAS Number: 270573-71-2
EINECS: 205-741-5

Order Now

Specification Test Results

Solubility 1350 g/l (20 °C)
Melting Point 300 °C
Molar Mass 332.22 g/mol
Bulk Density 465 kg/m3
pH 5.0 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR