Nitric acid 69%, EMPARTA® analytical reagent, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Nitric acid solution

M-Clarity™ quality level = MQ200

1.01832.2500
EM1.01832.2500CS 462 USD
EM1.01832.2500
Nitric acid 69%, EMPARTA® analytical reagent, Supelco®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Appearance passes test
As (Arsenic) ≤ 0.010 ppm
Assay (acidimetric) 68.0 - 70.0 %
Chloride (Cl) ≤ 0.5 ppm
Colour ≤ 10 Hazen
Fe (Iron) ≤ 0.200 ppm
Heavy metals (as Pb) ≤ 0.2 ppm
Identity passes test
Residue on ignition (as SO₄) ≤ 5 ppm
Sulphate (SO₄) ≤ 1 ppm
Sulfate (SO₄) ≤ 1 ppm
Titratable base ≤ 0.0004 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR