Acetic acid glacial ≥100%, EMPARTA® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.01830.2500
EM1.01830.2500CS 357 USD
EM1.01830.2500
Acetic acid glacial ≥100%, EMPARTA® ACS for analysis, Supelco®
Acetic acid
Formula: H₃CCOOH
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3),
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acetic anhydride ≤ 100 ppm
Assay (acidimetric) ≥ 99.7 %
Chloride (Cl) ≤ 1 ppm
Color ≤ 10 Hazen
Dilution test passes test
Evaporation residue ≤ 10 ppm
Fe (Iron) ≤ 0.2 ppm
Heavy metals (as Pb) ≤ 0.5 ppm
Substances reducing potassium dichromate passes test
Substances reducing potassium permanganate passes test
Sulfate (SO₄) ≤ 1 ppm
Titratable base ≤ 0.0004 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR