Test Kits for Spectroquant® Prove, NOVA 30/60, Move and Multy, MilliporeSigma
Supplier: MilliporeSigma
1.01812.0001
EM1.01812.0001CS 1615 USD
EM1.01812.0001
Test Kits for Spectroquant® Prove, NOVA 30/60, Move and Multy, MilliporeSigma
Environmental Test Kits
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments.
  • Barcode identification system of test kit by Spectroquant photometers enables rapid and simple operation
  • Preprogrammed blank values ensure fast reliable results
  • Validated standard- compliant reagents ensure secure analysis
Tests can be used not only on photometers and spectrophotometer from Merck Millipore, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
Spectroquant® cell test kits contain the necesary reagents for the analysis in the cells for most of the parameters. Packing design protects the cells from fluctuation of temperarture and moisture during storage and transport. The barcode on the cells ensures the correct method of analysis is chosen- elimiating mistakes and reducing set up times.
Spectroquant® reagent test kits is highly stable and ready-to-use reagent mixtures in a user friendly design. Free selection of appropriate cell format according to the measuring range. Long shelf life upto 3 years, if all are at room temperature. Automatic selection of the correct method with the help of Autoselector barcode identification.
Note: The range quoted in the table below refers to the Prove, and Nova instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Certifications: Complies with ISO 15705