Potassium Chloride Solution (Nominal 0.015 mS/cm) Certified Reference Material for the Measurement of electrolytic Conductivity, traceable to PTB and NIST (c=0.0001 mol/L), MilliporeSigma
Supplier: MilliporeSigma
1.01811.0105
EM1.01811.0105CS 3237 USD
EM1.01811.0105
Potassium Chloride Solution (Nominal 0.015 mS/cm) Certified Reference Material for the Measurement of electrolytic Conductivity, traceable to PTB and NIST (c=0.0001 mol/L), MilliporeSigma
Standards Conductivity Standards