Benzene-[D₆] ≥99.6% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.01789.0009 1.01789.0010 1.01789.0100
EM1.01789.0009EA 182 USD
EM1.01789.0009 EM1.01789.0010 EM1.01789.0100
Benzene-[D₆] ≥99.6% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Benzene-[D6]

MagniSolv™ deuterated solvents provide:
• Reliable results in the NMR-spectra by excellent chemical purity and highest isotopic enrichment.
• Reliable deuteration degrees.
• Low residual water content.
• Determination of water content in two ways.

Formula: C₆D₆
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: 6.7 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: –11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003010
CAS Number: 1076-43-3
EINECS: 214-061-8
UN: 1114
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR