Surfactants (cation.) Cell Test Method: photometric 0.05 - 1.50 mg/L CTAB Spectroquant®
Supplier: MilliporeSigma
1.01764.0001
EM1.01764.0001CS 1821 USD
EM1.01764.0001
Surfactants (cation.) Cell Test Method: photometric 0.05 - 1.50 mg/L CTAB Spectroquant®
Environmental Test Kits