Barium hydroxide octahydrate, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.01735.1000
EM1.01735.1000CS 4800 USD
EM1.01735.1000
Barium hydroxide octahydrate, Supelco®
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
Boiling Pt: 780 °C (1013 hPa)
Melting Pt: 78 °C
Density: 2.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 2923
ADR: 8 (6.1),II
Merck Index: 13,00980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR