Barium fluoride, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.01722.0050
EM1.01722.0050CS 52951 USD
EM1.01722.0050
Barium fluoride, Suprapur® for trace metal analysis, Supelco®
Barium fluoride
Formula: BaF₂
Boiling Pt: 2260 °C (1013 hPa)
Melting Pt: 1270 °C
Density: 4.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01003

Order Now

Specification Test Results

Solubility 1.2 g/l (25 °C)
Melting Point 1353 °C
Molar Mass 175.33 g/mol
Boiling Point 2260 °C
Density 4.89 g/cm3 (20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR