Barium acetate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.01704.0500
EM1.01704.0500CS 5434 USD
EM1.01704.0500
Barium acetate, EMSURE® for analysis, Supelco®
Barium acetate
Formula: (H₃CCOO)₂Ba
Melting Pt: ∼450 °C
Density: 2.468 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III
Merck Index: 13,00968

Order Now

Specification Test Results

Solubility 720 g/l (20 °C)
Melting Point 450 °C
Molar Mass 255.42 g/mol
Bulk Density 1200 kg/m3
Density 2.47 g/cm3 (20 °C)
pH 6.5 - 8.5 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR