Absorbent, Chemizorb® Hydrofluoric Acid, EMD Millipore
Supplier: MilliporeSigma
1.01591.1000
EM1.01591.1000CS 1396 USD
EM1.01591.1000
Absorbent, Chemizorb® Hydrofluoric Acid, EMD Millipore
Spill Control Materials
Chemizorb® Hydrofluoric Acid (HF) is a powder mixture consisting of a synthetic copolymer as the absorbent, a calcium salt as a neutraliser and precipitant, and a pH indicator. The neutralisation process results in the release of CO₂. When the hydrofluoric acid has been neutralised, the colour of the indicator changes from red to orange/yellow.
  • Easy to use
  • pH indicator makes it possible to monitor the neutralisation process
About 150 g of Chemizorb® HF is required to absorb and neutralise 100 ml of hydrofluoric acid.
Merck products are not available from Avantor in all countries. Please check with your local distributor.