Silver ≥99.9%, wool for elementary analysis, Supelco®

Supplier: MilliporeSigma

1.01506.0010
EM1.01506.0010CS 17695 USD
EM1.01506.0010
Silver ≥99.9%, wool for elementary analysis, Supelco®
Silver
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 961.9 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Specification Test Results

Identity passes test
Purity ≥ 99.9 %
Diameter of fibre about 45 - 55 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR