Anthrone, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Anthracen-9(10H)-one

1.01468.0010
EM1.01468.0010CS 5267 USD
EM1.01468.0010
Anthrone, GR for analysis, Supelco®
Anthrone
Formula: C₁₄H₁₀O
MW: 194.23 g/mol
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0

Order Now

Specification Test Results

Solubility 4.6 mg/l (20 °C)
Melting Point 151 - 156 °C
Molar Mass 194.23 g/mol
Bulk Density 450 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR