Ammonium dihydrogen phosphate, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

1.01440.0050
EM1.01440.0050CS 48828 USD
EM1.01440.0050
Ammonium dihydrogen phosphate, Suprapur® for trace metal analysis, Supelco®
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C
Density: 1.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547

Order Now

Specification Test Results

Solubility 368 g/l (20 °C)
Melting Point 190 °C
Molar Mass 115.02 g/mol
Bulk Density 800 - 1100 kg/m3
Vapor Pressure 0.066 hPa (125 °C)
Density 1.80 g/cm3 (20 °C)
pH 3.8 - 4.4 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR