Crystal violet indicator (C.I. 42555), Supelco®

Supplier: MilliporeSigma
Danger

1.01408.0025 1.01408.0100 1.01408.1000
EM1.01408.0025CS 1285 USD
EM1.01408.0025 EM1.01408.0100 EM1.01408.1000
Crystal violet indicator (C.I. 42555), Supelco®
Crystal Violet
Formula: C₂₅H₃₀ClN₃
Melting Pt: 189…194 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406

Order Now

Specification Test Results

Solubility 10 g/l (20 °C)
Melting Point 189 - 194 °C
Molar Mass 407.99 g/mol
Bulk Density 220 - 400 kg/m3
Density 1.19 g/cm3 (20 °C)
pH 2.5 - 3.5 (10 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR