Leishman's eosin-methylene blue stain for cytology, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Eosin and methylene blue Leishman, Eosin-Methylene blue dye Leishman, Leishman's stain, Leishman stain, Leishman's stain (eosin methylene blue compound)

M-Clarity™ quality level = MQ400

1.01350.0010
EM1.01350.0010CS 3444 USD
EM1.01350.0010
Leishman's eosin-methylene blue stain for cytology, Sigma-Aldrich®
Leishman's Eosin-Methylene blue

Used to stain blood, bone marrow smears, and clinical cytological specimens.


The dry mixture of the Leishman's eosin methylene blue dye is used for the self-preparation of the Leishman's staining solution. The staining method according to Leishman is one of the standard techniques in hematological diagnostic procedures. Like Wright's eosin methylene blue and May-Grünwald's eosin methylene blue, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The dye is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.

Storage Temperature: Ambient
MDL Number: MFCD00131498
CAS Number: 12627-53-1
EINECS: 235-732-1
IVDLogo

Order Now

Specification Test Results

Identity (UV/VIS-Spectrum) passes test
pH-value (0,01 %; methanol 50 %) 5 - 6
Absorption maximum λ 1 (Methanol) 623 - 627 nm
Absorption maximum λ 2 (Methanol) 522 - 525 nm
Spec. Absorptivity A 1%/1cm
- λ1max; 0.0005 %; methanol; calculated on dried substance 800 - 1100
- λ2max; 0.0005 %; methanol; calculated on dried substance 650 - 850
TLC-Test passes test
Loss on drying (105 °C) ≤ 8 %
Suitability for microscopy passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR