Ammonium thiocyanate ≥99.0% (silver nitrate titration), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.01213.0500 1.01213.9025
EM1.01213.0500CS 2617 USD
EM1.01213.0500 EM1.01213.9025
Ammonium thiocyanate ≥99.0% (silver nitrate titration), EMSURE® for analysis, Supelco®
Ammonium thiocyanate

1.01213.0500 replaces and is the same as AX1415-1. Part # Change only, Same Pack Size, Same Specifications.
1.01213.0500 replaces and is the same as AX1415-2 . Different Pack Size, Same Specifications.
1.01213.0500 replaces and is the same as AX1415-5. Different Pack Size, Same Specifications.

Formula: NH₄SCN
Melting Pt: 151 °C
Density: 1.3 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
UN: 3077
Merck Index: 13,00564

Order Now

Specification Test Results

Solubility 1600 g/l (20 °C)
Melting Point 150 °C
Molar Mass 76.12 g/mol
Bulk Density 600 - 700 kg/m3
Vapor Pressure  - 1 hPa (20 °C)
Density 1.3 g/cm3 (20 °C)
pH 4.8 - 5.8 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR