Ammonium persulfate (APS), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Peroxidisulfuric acid diammonium salt, Ammonium persulfate, Peroxidisulphuric acid diammonium salt, Ammonium persulphate

1.01201.0500 1.01201.1000 1.01201.5000 1.01201.9012
EM1.01201.0500CS 740 USD
EM1.01201.0500 EM1.01201.1000 EM1.01201.5000 EM1.01201.9012
Ammonium persulfate (APS), EMSURE® for analysis, Supelco®
Ammonium peroxodisulphate (APS)

1.01201.0500 replaces and is the same as AX1340-1. Part # Change only, Same Pack Size, Same Specifications.
1.01201.5000 replaces and is the same as AX1340-3. Different Pack Size, Same Specifications.
1.01201.9050 replaces and is the same as AX1340-20. Part # Change only, Same Pack Size, Same Specifications.

Formula: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Melting Pt: 120 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 620 g/l (20 °C)
Melting Point 120 °C (decomposition)
Molar Mass 228.19 g/mol
Bulk Density 900 kg/m3
Density 1.98 g/cm3 (20 °C)
pH 3.2 (100 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR