Ammonium nitrate, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.01187.5000
EM1.01187.5000EA 1023 USD
EM1.01187.5000
Ammonium nitrate, Supelco®
Ammonium nitrate
Formula: NH₄NO₃
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 169 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00011425
CAS Number: 6484-52-2
UN: 1942
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR