Ammonium heptamolybdate tetrahydrate, GR for analysis, Supelco®

Supplier: MilliporeSigma

1.01182.0250 1.01182.1000 1.01182.5000 1.01182.9025
EM1.01182.0250CS 1151 USD
EM1.01182.0250 EM1.01182.1000 EM1.01182.5000 EM1.01182.9025
Ammonium heptamolybdate tetrahydrate, GR for analysis, Supelco®
Ammonium heptamolybdate tetrahydrate

Order Now

Specification Test Results

Solubility 400 g/l (20 °C)
Melting Point 90 °C Elimination of water of crystallisation
Molar Mass 1235.86 g/mol
Bulk Density 800 kg/m3
Density 2.498 g/cm3 (20 °C)
pH 5.3 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR