Ammonium fluoride ≥98.0% (by precipitation titration), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Fluoroammonium

1.01164.0250 1.01164.1000 1.01164.9025
EM1.01164.0250CS 1865 USD
EM1.01164.0250 EM1.01164.1000 EM1.01164.9025
Ammonium fluoride ≥98.0% (by precipitation titration), EMSURE® for analysis, Supelco®
Ammonium fluoride

1.01164.0250 is offered as an alternative to AX1295-1. Different Pack Size, Same Specifications.
1.01164.1000 is offered as an alternative to AX1295-1. different size, different packaging, same chem spec.

Formula: NH₄F
MW: 37.04 g/mol
Melting Pt: 160
Density: 1.01 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1 III
Merck Index: 13,00524

Order Now

Specification Test Results

Solubility 820 g/l (20 °C)
Molar Mass 37.04 g/mol
Bulk Density 250 - 350 kg/m3
Density 1.01 g/cm3 (20 °C)
pH 6 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR