Ammonium dihydrogen phosphate, Supelco®

Supplier: MilliporeSigma

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

M-Clarity™ quality level = MQ300

1.01126.5000
EM1.01126.5000EA 1424 USD
EM1.01126.5000
Ammonium dihydrogen phosphate, Supelco®
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C
Density: 1.8 g/cm³ (20 °C)
MDL Number: MFCD00003396
CAS Number: 7722-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR