Aluminium Oxide 60 GF₂₅₄ Neutral (type E) for Thin-Layer Chromatography, Merck
Supplier: MilliporeSigma
1.01092.0500
EM1.01092.0500EA 228.66 USD
EM1.01092.0500
Aluminium Oxide 60 GF₂₅₄ Neutral (type E) for Thin-Layer Chromatography, Merck
Chromatography Sorbents TLC Sorbents
Standardized sorbents ensure reliable results every time
EMD Millipore's broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µ: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).