Aluminium hydroxide (hydrargillite), powder, EMPLURA®, Supelco®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
1.01091.1000
1.01091.9050
EM1.01091.1000CS
660
USD
EM1.01091.1000
EM1.01091.9050
Aluminium hydroxide (hydrargillite), powder, EMPLURA®, Supelco®
Aluminium hydroxide
Formula:
Al(OH)₃ Melting Pt: 300 °C (–H₂O of crystall.) Density: 2.42 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003420 CAS Number: 21645-51-2 EINECS: 244-492-7 Merck Index: 13,00342 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...