1-Propanol, LiChrosolv® for HPLC, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.01024.1000 1.01024.2500
EM1.01024.1000CS 1203 USD
EM1.01024.1000 EM1.01024.2500
1-Propanol, LiChrosolv® for HPLC, Supelco®
1-Propanol
Formula: H₃CCH₂CH₂OH
Boiling Pt: 97 °C (1013 hPa)
Melting Pt: –127 °C
Density: 0.8053 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99.8 %
Identity (IR) conforms
Evaporation residue ≤ 2.0 mg/l
Water ≤ 0.02 %
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Transmission (at 230 nm) ≥ 70 %
Transmission (at 240 nm) ≥ 80 %
Transmission (from 270 nm) ≥ 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR