2-Propanol ≥99.5% (by GC), EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®

Supplier: MilliporeSigma
Danger

1.00995.6190
EM1.00995.4003EA 622.9 USD
EM1.00995.4003 EM1.00995.6190
2-Propanol ≥99.5% (by GC), EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
2-Propanol

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certifications: Conforms to Ph. Eur., BP, JP, USP

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) ≥ 99.5 %
Identity (IR) conforms
Identity B (USP) conforms
Identity (JP) conforms
Appearance conforms
Clarity of solution conforms
Acidity or alkalinity conforms
Acidity conforms
Refractive index (n 20/D) 1.376 - 1.378
Density (d 20 °C/20 °C) 0.785 - 0.788
Density (d 25 °C/25 °C) 0.783 - 0.787
Boiling range (81-83°C) ≥ 94 % (v/v)
Absorbance (at 230 nm) ≤ 0.300
Absorbance (at 250 nm) ≤ 0.100
Absorbance (at 270 nm) ≤ 0.030
Absorbance (at 290 nm) ≤ 0.020
Absorbance (at 310 nm) ≤ 0.010
The spectrum shows a steadily descending curve with no observable peaks or shoulders conforms
Peroxides conforms
Volatile Impurities (GC) (individual) ≤ 0.1 % (v/v)
Volatile Impurities (GC) (total) ≤ 1.0 % (v/v)
Benzene and related substances (GC) (Benzene) ≤ 2 ppm
Benzene and related substances (GC) (total of impurities apart from 2-butanol) ≤ 0.3 %
Cyclohexane (GC) ≤ 50 ppm
Acetone (GC) ≤ 50 ppm
Ethanol (GC) ≤ 50 ppm
1-Propylalcohol (GC) ≤ 50 ppm
Isopropylether (GC) ≤ 0.01 %
Other residual solvents (ICH Q3C) excluded by manufacturing process
Evaporation residue ≤ 0.0020 %
Water ≤ 0.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR