Isobutanol ≥98.5% (by GC), Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.00985.2500 1.00985.6025 1.00985.9190
EM1.00985.2500CS 733 USD
EM1.00985.2500 EM1.00985.6025 EM1.00985.9190
Isobutanol ≥98.5% (by GC), Supelco®
Isobutanol
Formula: (CH₃)₂CHCH₂OH
Boiling Pt: 106…108 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.802 g/cm3 (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR