Ethanol denatured ≥99.5% (by GC), EMSURE® for analysis (denatured with 1% MEK), Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ethyl alcohol denatured

M-Clarity™ quality level = MQ300

1.00974.1011 1.00974.2511 1.00974.6025 1.00974.9025 1.00974.9180 1.00974.2500
EM1.00974.1011CS 650 USD
EM1.00974.1011 EM1.00974.2511 EM1.00974.6025 EM1.00974.9025 EM1.00974.9180 EM1.00974.2500
Ethanol denatured ≥99.5% (by GC), EMSURE® for analysis (denatured with 1% MEK), Supelco®
Ethanol denatured
Formula: H₃CCH₂OH
MW: 46.07 g/mol
Melting Pt: –114.5 °C (ethanol)
Density: 0.79 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II

Order Now

Specification Test Results

Purity (GC) ≥ 99.5 %
Identity (IR) conforms
Free acid (as acetic acid) ≤ 0.001 %
Ethylmethylketone (GC) 0.8 - 1.3 %
Readily carbonizable substances conforms
Evaporation residue ≤ 0.001 %
Water ≤ 0.1 %
Al (Aluminium) ≤ 0.00005 %
B (Boron) ≤ 0.00005 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
K (Potassium) ≤ 0.0001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR