Tetrachloroethylene ≥99% (by GC), Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.00964.1000 1.00964.2500 1.00964.6025 1.00964.9190
EM1.00964.1000CS 837 USD
EM1.00964.1000 EM1.00964.2500 EM1.00964.6025 EM1.00964.9190
Tetrachloroethylene ≥99% (by GC), Supelco®
Tetrachloroethylene
Formula: C₂Cl₄
Boiling Pt: 121 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.62 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III
Merck Index: 12,09332
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR