Diethyl ether, SupraSolv® for GC-ECD/FID, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane, Ethyl ether

1.00931.2500
EM1.00931.2500CS 1609 USD
EM1.00931.2500
Diethyl ether, SupraSolv® for GC-ECD/FID, Supelco®
Diethyl ether

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 69 g/l (20 °C)
Melting Point -116.3 °C
Molar Mass 74.12 g/mol
Boiling Point 34.6 °C (1013 hPa)
Vapor Pressure 587 hPa (20 °C)
Flash Point -40 °C
Explosion Limit 1.7 - 36 %(V)
Density 0.71 g/cm3
Ignition Point 180 °C
Saturation concentration 1776 g/m3 (20 °C) Air
Evaporation number 1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR