Entellan® Mounting Medium, MilliporeSigma
Supplier: MilliporeSigma
1.00869.0500
EM1.00869.0500EA 135 USD
EM1.00869.0500
Entellan® Mounting Medium, MilliporeSigma
Stains Microscopy and staining reagents
Entellan® new for cover slippers is a new water-free mounting medium suited for standard, commercial automatic mounting instruments that operate with cover slippers
Its viscosity range is adjusted to a more narrow range than other mounting media, which minimizes the effort for the re-calibration of the instrument when a new bottle is used. As the mounting medium is an IVD product and CE registered it can be used in diagnostics and for laboratory accreditations.