Supelco® Spectroquant® Rectangular cells quartz 10 mm, EMD Millipore
Supplier: MilliporeSigma
1.00784.0001
EM1.00784.0001EA 777.72 USD
EM1.00784.0001
Supelco® Spectroquant® Rectangular cells quartz 10 mm, EMD Millipore
Cuvettes Spectrophotometer Cuvettes
Cuvettes made of quartz glas are especially suitable to perform measurements using a wavelength from the UV range. In contrast to other cuvettes, these cells don't absorb in this range and therefore allow for safe measurement results. They can be used in combination with photometes with a cell holder for 10 mm rectangular cells, e.g. the Spectroquant Prove series.
Storage temperature: 2…30 °C