Mercaptoacetic acid (thioglycolic acid), GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Mercaptoacetic acid

M-Clarity™ quality level = MQ100

1.00700.1000
EM1.00700.0100CS 2261.75 USD
EM1.00700.0100 EM1.00700.1000
Mercaptoacetic acid (thioglycolic acid), GR for analysis, Supelco®
Mercaptoacetic acid (Thioglycolic acid)
Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 96
Melting Pt: -16.5
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 130 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
Merck Index: 13,09410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45179825145ec44f2e058197a0ee54845ae7c750&vid=51e43f5b69b880b196570b3065f9b4d8db7336d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Description clear, colourless liquid
Assay (iodometric) ≥ 99.0 %
Solubility (USP) conforms
Solubility in water conforms
Solubility in ethanol conforms
Sensitivity (USP) conforms
Fe (Iron) ≤ 0.00005 %
Organic bound chlorine ≤ 0.01 %
Sulfated ash ≤ 0.05 %
Identity (IR) conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR