Ammonium Test, Spectroquant®, MilliporeSigma
Supplier: MilliporeSigma
1.14752.0001 1.14752.0002 1.00683.0001
EM1.14752.0001CS 2960 USD
EM1.14752.0001 EM1.14752.0002 EM1.00683.0001
Ammonium Test, Spectroquant®, MilliporeSigma
Environmental Test Kits
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).