Succinic acid, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: 1,4-Butanedioic acid

1.00682.0250
EM1.00682.0250CS 1244 USD
EM1.00682.0250
Succinic acid, EMSURE® for analysis, Supelco®
Succinic acid

In North America 1.00682.0250 is similar to SX1040-3 but in a smaller pack size

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 182 °C
Density: 1.56 g/cm³ (20 °C)
Flash Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8,III
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 58 g/l (20 °C)
Melting Point 185 - 190 °C
Molar Mass 118.09 g/mol
Bulk Density 940 kg/m3
Boiling Point 235 °C (1013 hPa)
Flash Point 206 °C
Density 1.56 g/cm3 (20 °C)
pH 2.7 (10 g/l, H2O, 20 °C)
Ignition Point 470 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR