Tungstosilicic acid hydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

1.00659.0100
EM1.00659.0100CS 16138 USD
EM1.00659.0100
Tungstosilicic acid hydrate, EMSURE® for analysis, Supelco®
Tungstosilicic acid Hydrate
Formula: H₄Si(W₃O₁₀)₄.xH₂O
MDL Number: MFCD00036271
CAS Number: 12027-43-9

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 2878.17 g/mol
Bulk Density 200 - 500 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR