Water, SupraSolv® for headspace gas chromatography, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

1.00577.1000 1.00577.2500
EM1.00577.1000CS 817 USD
EM1.00577.1000 EM1.00577.2500
Water, SupraSolv® for headspace gas chromatography, Supelco®
Water

SupraSolv® solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur, and USP - SupraSolv® headspace solvents are tested for all residual solvents defined by the ICH.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Molar Mass 18.02 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 23 hPa (20 °C)
Density 1.00 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR