Tryptic Soy Broth, Non-Animal, Irradiated, Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
1.00550.0500
EM1.00550.0500EA 242.34 USD
EM1.00550.0500
Tryptic Soy Broth, Non-Animal, Irradiated, Dehydrated Culture Media, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Media is granulated.
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.